Assay Details
| Gene Symbol: cst-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029404.7 | NP_001024575.2 | 9 - 10 | 1293 | 84 | |
| NM_076342.6 | NP_508743.4 | 9 - 10 | 1285 | 84 | ||
| GenBank mRNA | AF271359.1 | - | 9 - 10 | 1281 | 84 | |
More Information
Set Membership: |
|

cst-1
Genomic Map