Assay Details
| Gene Symbol: mltn-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076364.4 | NP_508765.2 | 10 - 11 | 1905 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mltn-13| Gene Symbol: mltn-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076364.4 | NP_508765.2 | 10 - 11 | 1905 | 81 | |
Set Membership: |
|