Assay Details
| Gene Symbol: dgk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029508.6 | NP_001024679.1 | 10 - 11 | 1600 | 98 | |
| NM_001383562.1 | 10 - 11 | 1596 | 98 | |||
More Information
Set Membership: |
|

dgk-2
Genomic Map