Assay Details
| Gene Symbol: dgk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029508.6 | NP_001024679.1 | 4 - 5 | 716 | 104 | |
| NM_001383562.1 | 4 - 5 | 712 | 104 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383562.1 transcripts.
Set Membership: |
|

dgk-2
Genomic Map