Assay Details
| Gene Symbol: dgk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029508.6 | NP_001024679.1 | 5 - 6 | 850 | 90 | |
| NM_001029509.5 | NP_001024680.1 | 1 - 2 | 135 | 90 | ||
| NM_001383562.1 | 5 - 6 | 846 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001029509.5, NM_001383562.1 transcripts.
Set Membership: |
|

dgk-2
Genomic Map