Assay Details
| Gene Symbol: dgk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029508.6 | NP_001024679.1 | 9 - 10 | 1434 | 104 | |
| NM_001029509.5 | NP_001024680.1 | 4 - 5 | 762 | 104 | ||
| NM_001383562.1 | 9 - 10 | 1430 | 104 | |||
More Information
Set Membership: |
|

dgk-2
Genomic Map