Assay Details
| Gene Symbol: ham-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029192.6 | NP_001024363.2 | 4 - 5 | 792 | 98 | |
| NM_001029193.7 | NP_001024364.2 | 4 - 5 | 797 | 98 | ||
| NM_001307898.3 | NP_001294827.1 | 3 - 4 | 605 | 98 | ||
More Information
Set Membership: |
|

ham-2
Genomic Map