Assay Details
| Gene Symbol: mfsd-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076398.6 | NP_508799.1 | 6 - 7 | 559 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mfsd-12| Gene Symbol: mfsd-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076398.6 | NP_508799.1 | 6 - 7 | 559 | 83 | |
Set Membership: |
|