Assay Details
| Gene Symbol: mls-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076414.7 | NP_508815.2 | 5 - 6 | 892 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mls-2| Gene Symbol: mls-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076414.7 | NP_508815.2 | 5 - 6 | 892 | 103 | |
Set Membership: |
|