Assay Details
| Gene Symbol: add-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029469.5 | NP_001024640.1 | 15 - 16 | 2004 | 73 | |
| NM_001380952.1 | NP_001367832.1 | 15 - 16 | 1997 | 73 | ||
More Information
Set Membership: |
|

add-1
Genomic Map