Assay Details
| Gene Symbol: add-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029468.5 | NP_001024639.1 | 8 - 9 | 914 | 77 | |
| NM_001029469.5 | NP_001024640.1 | 8 - 9 | 921 | 77 | ||
| NM_001029470.4 | NP_001024641.1 | 8 - 9 | 923 | 77 | ||
| NM_001029471.2 | NP_001024642.1 | 8 - 9 | 914 | 77 | ||
| NM_001380952.1 | NP_001367832.1 | 8 - 9 | 914 | 77 | ||
More Information
Set Membership: |
|

add-1
Genomic Map