Assay Details
| Gene Symbol: calu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383568.1 | 3 - 4 | 532 | 74 | ||
| NM_001383569.1 | 1 - 2 | 109 | 74 | |||
| GenBank mRNA | AF228528.1 | - | 1 - 2 | 122 | 74 | |
More Information
Set Membership: |
|

calu-1
Genomic Map