Assay Details
| Gene Symbol: calu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383568.1 | 4 - 5 | 645 | 88 | ||
| GenBank mRNA | AF228528.1 | - | 2 - 3 | 235 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

calu-1| Gene Symbol: calu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383568.1 | 4 - 5 | 645 | 88 | ||
| GenBank mRNA | AF228528.1 | - | 2 - 3 | 235 | 88 | |
Set Membership: |
|