Assay Details
| Gene Symbol: dpy-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076472.4 | NP_508873.1 | 3 - 4 | 1279 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-8| Gene Symbol: dpy-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076472.4 | NP_508873.1 | 3 - 4 | 1279 | 75 | |
Set Membership: |
|