Assay Details
| Gene Symbol: mtm-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076487.4 | NP_508888.2 | 3 - 4 | 586 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtm-5| Gene Symbol: mtm-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076487.4 | NP_508888.2 | 3 - 4 | 586 | 83 | |
Set Membership: |
|