Assay Details
| Gene Symbol: vps-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380962.1 | NP_001367568.1 | 9 - 10 | 2103 | 97 | |
| NM_001383575.1 | 9 - 10 | 2108 | 97 | |||
| NM_001383576.1 | 7 - 8 | 1677 | 97 | |||
More Information
Set Membership: |
|

vps-41
Genomic Map