Assay Details
| Gene Symbol: vps-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380962.1 | NP_001367568.1 | 3 - 4 | 542 | 140 | |
| NM_001383575.1 | 3 - 4 | 547 | 140 | |||
| NM_001383576.1 | 1 - 2 | 116 | 140 | |||
More Information
Set Membership: |
|

vps-41
Genomic Map