Assay Details
| Gene Symbol: pfn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076509.6 | NP_508910.3 | 1 - 2 | 144 | 83 | |
| GenBank mRNA | AY530909.1 | - | 1 - 2 | 126 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pfn-2| Gene Symbol: pfn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076509.6 | NP_508910.3 | 1 - 2 | 144 | 83 | |
| GenBank mRNA | AY530909.1 | - | 1 - 2 | 126 | 83 | |
Set Membership: |
|