Assay Details
| Gene Symbol: pks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330898.4 | NP_001317900.1 | 24 - 25 | 12803 | 83 | |
| NM_076522.3 | NP_508923.2 | 23 - 24 | 22307 | 83 | ||
More Information
Set Membership: |
|

pks-1
Genomic Map