Assay Details
| Gene Symbol: pks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330898.4 | NP_001317900.1 | 26 - 27 | 13226 | 79 | |
| NM_076522.3 | NP_508923.2 | 25 - 26 | 22730 | 79 | ||
More Information
Set Membership: |
|

pks-1
Genomic Map