Assay Details
| Gene Symbol: ddr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076525.5 | NP_508926.2 | 3 - 4 | 424 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddr-1| Gene Symbol: ddr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076525.5 | NP_508926.2 | 3 - 4 | 424 | 103 | |
Set Membership: |
|