Assay Details
| Gene Symbol: dlg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029260.4 | NP_001024431.1 | 6 - 7 | 1164 | 67 | |
| GenBank mRNA | AF406786.1 | - | 6 - 7 | 1103 | 67 | |
| AJ295228.1 | - | 6 - 7 | 1171 | 67 | ||
More Information
Set Membership: |
|

dlg-1
Genomic Map