Assay Details
| Gene Symbol: ctbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270101.4 | NP_001257030.1 | 10 - 11 | 1407 | 107 | |
| NM_001270102.3 | NP_001257031.1 | 7 - 8 | 1044 | 107 | ||
More Information
Set Membership: |
|

ctbp-1
Genomic Map