Assay Details
| Gene Symbol: ctbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270101.4 | NP_001257030.1 | 11 - 12 | 1513 | 86 | |
| NM_001270102.3 | NP_001257031.1 | 8 - 9 | 1150 | 86 | ||
More Information
Set Membership: |
|

ctbp-1
Genomic Map