Assay Details
| Gene Symbol: ctbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270101.4 | NP_001257030.1 | 7 - 8 | 844 | 93 | |
| NM_001270102.3 | NP_001257031.1 | 4 - 5 | 481 | 93 | ||
More Information
Set Membership: |
|

ctbp-1
Genomic Map