Assay Details
| Gene Symbol: flp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076584.6 | NP_508985.1 | 1 - 2 | 98 | 76 | |
| GenBank mRNA | AF042393.1 | - | 1 - 2 | 98 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-7| Gene Symbol: flp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076584.6 | NP_508985.1 | 1 - 2 | 98 | 76 | |
| GenBank mRNA | AF042393.1 | - | 1 - 2 | 98 | 76 | |
Set Membership: |
|