Assay Details
| Gene Symbol: flp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076584.6 | NP_508985.1 | 3 - 4 | 429 | 103 | |
| GenBank mRNA | AF042393.1 | - | 3 - 4 | 429 | 103 | |
More Information
Set Membership: |
|

flp-7
Genomic Map