Assay Details
| Gene Symbol: kin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029671.6 | NP_001024842.1 | 2 - 3 | 303 | 78 | |
| NM_001083302.4 | NP_001076771.1 | 3 - 4 | 407 | 78 | ||
| NM_001380985.1 | NP_001367669.1 | 3 - 4 | 391 | 78 | ||
More Information
Set Membership: |
|

kin-2
Genomic Map