Assay Details
| Gene Symbol: kin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029671.6 | NP_001024842.1 | 4 - 5 | 541 | 77 | |
| NM_001083302.4 | NP_001076771.1 | 5 - 6 | 645 | 77 | ||
| NM_001380985.1 | NP_001367669.1 | 5 - 6 | 629 | 77 | ||
More Information
Set Membership: |
|

kin-2
Genomic Map