Assay Details
| Gene Symbol: prx-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129639.3 | NP_001123111.1 | 7 - 8 | 908 | 133 | |
| NM_001383596.1 | 6 - 7 | 973 | 133 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prx-3| Gene Symbol: prx-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129639.3 | NP_001123111.1 | 7 - 8 | 908 | 133 | |
| NM_001383596.1 | 6 - 7 | 973 | 133 | |||
Set Membership: |
|