Assay Details
Gene Symbol: prx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129639.3 | NP_001123111.1 | 2 - 3 | 155 | 122 | |
NM_001383596.1 | 2 - 3 | 159 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383596.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
