Assay Details
| Gene Symbol: nnt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076627.6 | NP_509028.1 | 7 - 8 | 1312 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nnt-1| Gene Symbol: nnt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076627.6 | NP_509028.1 | 7 - 8 | 1312 | 107 | |
Set Membership: |
|