Assay Details
| Gene Symbol: lrch-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076641.6 | NP_509042.1 | 3 - 4 | 458 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lrch-1| Gene Symbol: lrch-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076641.6 | NP_509042.1 | 3 - 4 | 458 | 77 | |
Set Membership: |
|