Assay Details
| Gene Symbol: mup-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076675.9 | NP_509076.1 | 5 - 6 | 807 | 63 | |
| GenBank mRNA | U44759.1 | - | 5 - 6 | 903 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mup-2| Gene Symbol: mup-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076675.9 | NP_509076.1 | 5 - 6 | 807 | 63 | |
| GenBank mRNA | U44759.1 | - | 5 - 6 | 903 | 63 | |
Set Membership: |
|