Assay Details
| Gene Symbol: cdf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313388.5 | NP_001300317.1 | 7 | 1667 | 108 | |
| NM_076694.7 | NP_509095.1 | 6 - 7 | 1530 | 108 | ||
| NM_076695.6 | NP_509096.1 | 8 - 9 | 1860 | 108 | ||
More Information
Set Membership: |
|

cdf-1
Genomic Map