Assay Details
| Gene Symbol: cdf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313388.5 | NP_001300317.1 | 3 | 316 | 84 | |
| NM_076695.6 | NP_509096.1 | 3 - 4 | 509 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdf-1| Gene Symbol: cdf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313388.5 | NP_001300317.1 | 3 | 316 | 84 | |
| NM_076695.6 | NP_509096.1 | 3 - 4 | 509 | 84 | ||
Set Membership: |
|