Assay Details
| Gene Symbol: smf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029621.5 | NP_001024792.2 | 10 - 11 | 1572 | 74 | |
| NR_132544.1 | 9 - 10 | 1319 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smf-1| Gene Symbol: smf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029621.5 | NP_001024792.2 | 10 - 11 | 1572 | 74 | |
| NR_132544.1 | 9 - 10 | 1319 | 74 | |||
Set Membership: |
|