Assay Details
| Gene Symbol: smf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029621.5 | NP_001024792.2 | 4 - 5 | 662 | 132 | |
| NR_132544.1 | 4 - 5 | 639 | 132 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smf-1| Gene Symbol: smf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029621.5 | NP_001024792.2 | 4 - 5 | 662 | 132 | |
| NR_132544.1 | 4 - 5 | 639 | 132 | |||
Set Membership: |
|