Assay Details
| Gene Symbol: slc-25A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076732.6 | NP_509133.1 | 1 - 2 | 105 | 118 | |
| GenBank mRNA | X76114.1 | - | 110 | 118 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc-25A10| Gene Symbol: slc-25A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076732.6 | NP_509133.1 | 1 - 2 | 105 | 118 | |
| GenBank mRNA | X76114.1 | - | 110 | 118 | ||
Set Membership: |
|