Assay Details
| Gene Symbol: atg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076744.9 | NP_509145.3 | 2 - 3 | 280 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atg-2| Gene Symbol: atg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076744.9 | NP_509145.3 | 2 - 3 | 280 | 153 | |
Set Membership: |
|