Assay Details
| Gene Symbol: alh-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076802.5 | NP_509203.1 | 1 - 2 | 295 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alh-10| Gene Symbol: alh-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076802.5 | NP_509203.1 | 1 - 2 | 295 | 84 | |
Set Membership: |
|