Assay Details
| Gene Symbol: alh-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076802.5 | NP_509203.1 | 8 - 9 | 1457 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alh-10| Gene Symbol: alh-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076802.5 | NP_509203.1 | 8 - 9 | 1457 | 58 | |
Set Membership: |
|