Assay Details
| Gene Symbol: lam-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076803.7 | NP_509204.3 | 9 - 10 | 3370 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lam-2| Gene Symbol: lam-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076803.7 | NP_509204.3 | 9 - 10 | 3370 | 101 | |
Set Membership: |
|