Assay Details
| Gene Symbol: pgp-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076804.5 | NP_509205.2 | 8 - 9 | 1029 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgp-10| Gene Symbol: pgp-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076804.5 | NP_509205.2 | 8 - 9 | 1029 | 75 | |
Set Membership: |
|