Assay Details
| Gene Symbol: syd-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375059.1 | NP_001362162.1 | 1 - 2 | 506 | 106 | |
| NM_001383605.1 | 1 - 2 | 471 | 106 | |||
| NM_076812.7 | NP_509213.1 | 1 - 2 | 145 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383605.1 transcripts.
Set Membership: |
|

syd-9
Genomic Map