Assay Details
| Gene Symbol: C07D8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076841.7 | NP_509242.1 | 4 - 5 | 972 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C07D8.6| Gene Symbol: C07D8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076841.7 | NP_509242.1 | 4 - 5 | 972 | 62 | |
Set Membership: |
|