Assay Details
| Gene Symbol: clc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375068.3 | NP_001362078.1 | 2 | 101 | 94 | |
| NM_076856.6 | NP_509257.1 | 1 - 2 | 94 | 94 | ||
| GenBank mRNA | AF252604.1 | - | 1 - 2 | 96 | 94 | |
More Information
Set Membership: |
|

clc-2
Genomic Map