Assay Details
| Gene Symbol: clc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076856.6 | NP_509257.1 | 4 - 5 | 395 | 86 | |
| GenBank mRNA | AF252604.1 | - | 4 - 5 | 397 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

clc-2| Gene Symbol: clc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076856.6 | NP_509257.1 | 4 - 5 | 395 | 86 | |
| GenBank mRNA | AF252604.1 | - | 4 - 5 | 397 | 86 | |
Set Membership: |
|