Assay Details
| Gene Symbol: sto-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129652.6 | NP_001123124.1 | 3 - 4 | 373 | 67 | |
| NM_076880.8 | NP_509281.1 | 3 - 4 | 409 | 67 | ||
| Gene Symbol: F08C6.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076879.5 | NP_509280.2 | 7 - 8 | 2585 | 67 | |
More Information
Set Membership: |
|

sto-1
Genomic Map