Assay Details
| Gene Symbol: sto-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129652.6 | NP_001123124.1 | 4 - 5 | 458 | 65 | |
| NM_076880.8 | NP_509281.1 | 4 - 5 | 494 | 65 | ||
More Information
Set Membership: |
|

sto-1
Genomic Map